missing translation for 'onlineSavingsMsg'
Learn More

LIMD2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 15471-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16844664

  • 3828.53 NOK / 150µL
Estimated Shipment: 17-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Acts as an activator of the protein-kinase ILK, thereby regulating cell motility.
Specifications

Specifications

LIMD2
Polyclonal
Unconjugated
LIMD2
LIM domain containing 2, LIMD2
Rabbit
Antigen Affinity Chromatography
RUO
360646, 67803, 80774
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q4KM31, Q8BGB5, Q9BT23
Limd2
LIMD2 Fusion Protein Ag7775
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers