Learn More
Cole-Parmer™ Lightnin High-Torque Direct-Drive Mixer
Use Lightnin High-Torque Direct-Drive Mixers in a wide range of applications. Choose a direct-drive model for small batch mixing or high-shear applications.
Brand:  Cole-Parmer™ 50310-36
Additional Details : Weight : 19.00000kg
 View more versions of this product
Product Code. 15447453
  • 28505.00 NOK / Each
Estimated Shipment: 23-08-2024
Description
  • Large selection lets you match mixer capability to your specific application
    • Mix up to 50 gallons of fluid.
    • These mixers offer more horsepower and torque than the small batch mixers
Specifications
Specifications
1750 rpm
0.75 in.
1/2 hp
316 Stainless Steel (SS) Shaft, 316 SS A-310 Impeller, and a Drive Quill with Set Screw.
EV1P25T1C48
140 in. oz.
208/230 V
19.05 mm
High-Torque Direct-Drive Mixer
316 Stainless Steel
TEFC
Direct-Drive Mixer