Learn More
Cole-Parmer™ Lightnin Heavy-Duty Gear Drive Mixer
Product Code. 15457443 Shop All Cole-Parmer Products
Change view
Click to view available options
Speed:
20 to 280 rpm
20 to 350 rpm
Model:
EV5P25A1D36
EV5P25A1F48
EV5P25A1J60
EV5P25ET1F48
EV5P25M1E36
EV5P25M1F48
EV5P25M1J60
EV5P25T1F48
EV5P25T1J60
EV5P50A2L36
EV5P50M2L36
EV5P50T2L36
EV5P5OAIP60
EV6P75M2X60
EV6P75T2T48
EV6P75T2X60
Motor Type:
Air-Driven
TEFC
TENV
XPRF
Torque:
1440 in. oz.
2701 in. oz.
720 in. oz.
Unit Size:
Each
Product Code. Speed Model Motor Type Torque unitSize
15457443 20 to 350 rpm EV5P25M1F48 TENV 720 in. oz. Each
15427453 20 to 280 rpm EV6P75M2X60 TENV 2701 in. oz. Each
15407463 20 to 280 rpm EV6P75T2T48 TEFC 2701 in. oz. Each
15417463 20 to 280 rpm EV6P75T2X60 TEFC 2701 in. oz. Each
15417443 20 to 350 rpm EV5P25M1E36 TENV 720 in. oz. Each
15427443 20 to 350 rpm EV5P25A1D36 Air-Driven 720 in. oz. Each
15467443 20 to 350 rpm EV5P25A1F48 Air-Driven 720 in. oz. Each
15407453 20 to 350 rpm EV5P25M1J60 TENV 720 in. oz. Each
15417453 20 to 350 rpm EV5P25A1J60 Air-Driven 720 in. oz. Each
15467453 20 to 350 rpm EV5P25ET1F48 XPRF 720 in. oz. Each
15477453 20 to 350 rpm EV5P25T1F48 TEFC 720 in. oz. Each
15487453 20 to 350 rpm EV5P25T1J60 TEFC 720 in. oz. Each
15437443 20 to 350 rpm EV5P50M2L36 TENV 1440 in. oz. Each
15447443 20 to 350 rpm EV5P50A2L36 Air-Driven 1440 in. oz. Each
15457453 20 to 350 rpm EV5P50T2L36 TEFC 1440 in. oz. Each
15437473 20 to 350 rpm EV5P5OAIP60 Air-Driven 1440 in. oz. Each
16 options
Product Code. 15457443
Brand: Cole-Parmer™ 5031008
Only null left
Add to basket
Add to basket
Lightnin Heavy-Duty Gear Drive Mixers are ideal for mixing your high-viscosity materials or large batches of water-like fluids. All models maintain speeds from 20 to 350 rpm (20:1 ratio) and generate enough torque to mix highly viscous fluids.
  • Includes:
    • 316 SS shaft, 316 SS A-310 impeller, and drive quill with set screw.
    • Models 50310-68, -70 and -72 also include bung adapter and a folding impeller.
  • More About This Item:
    • These variable-speed, gear-drive mixers are ideal for mixing your high-viscosity materials or large batches of water-like fluids. All models maintain speeds from 20 to 350 rpm (20:1 ratio) and generate enough torque to mix highly viscous fluids.
    • Gear-drive mixers mount with 20°> offset tank clamp (included). Available as either an air-powered or electric mixer.
    • Select from three speed options; 20 to 350 rpm (20:1 ratio), 20 to 280 rpm (14:1 ratio), or fixed speed of 350 or 280 rpm. The 3⁄4-hp mixers generate plenty of torque for your high-viscosity applications.
Specifications
Speed 20 to 350 rpm
Diameter (English) Shaft 0.75 in.
Diameter (Metric) Shaft 19.05 mm
Horsepower 1/4 hp
Includes 316 SS Shaft, 316 SS A-310 Impeller, and Drive Quill with Set Screw
Material 316 Stainless Steel
Model EV5P25M1F48
Motor Type TENV
Torque 720 in. oz.