Learn More
Hirschmann™ Light Protection Window
Product Code. 11652239
Change view
Click to view available options
For Use With (Equipment):
opus™
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
11652239 opus™ Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11652239 Supplier Hirschmann™ Supplier No. 9586060
  • ONLINE EXCLUSIVE PRICE
    288.75 NOK
    180.60 NOK / Pack of 1
    Save 108.15 NOK
    37% Off
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
Amber colored protection window for UV-sensitive media, used with opus™ and solarus™.
  • Amber colored replacement observation window
  • For the protection of UV-sensitive media
Specifications
Description For light sensitive media
Type UV Protection Window
Includes 1
For Use With (Equipment) opus™