Learn More
JULABO™ Lift-Up Bath Cover for PURA™ Water Baths
Product Code. p-7161038
Change view
Click to view available options
For Use With (Equipment):
PURA™ 10
PURA™ 14
PURA™ 22
PURA™ 30
PURA™ 4
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
16266611 PURA™ 4 Each
16276611 PURA™ 10 Each
16286611 PURA™ 14 Each
16296611 PURA™ 22 Each
16206621 PURA™ 30 Each
5 options
Product Code. 16266611
Brand: JULABO™ 9970580
Only null left
Add to basket
Add to basket
Lift-up, transparent bath cover for PURA™ Water Bath up to 99.9°C. Protects against water loss through evaporation. The bath cover is made from plexiglass and polycarbonate.
  • Accessory for PURA™ Water Bath
  • Prevent water loss through evaporation
  • Protect samples from contamination
  • Made from plexiglass and polycarbonate
Specifications
Description Lift-up bath cover
For Use With (Equipment) PURA™ 4
Material Plexiglass and polycarbonate