Learn More
JULABO™ Lift-Up Bath Cover for PURA™ Water Baths
Description
- Accessory for PURA™ Water Bath
- Prevent water loss through evaporation
- Protect samples from contamination
- Made from plexiglass and polycarbonate
Specifications
Specifications
| Description | Lift-up bath cover |
| For Use With (Equipment) | PURA™ 14 |
| Material | Plexiglass and polycarbonate |