Learn More
Lift Pump with Downtube
Click to view available options
Material:
Nylon
Unit Size:
Each
Description
For 20L/25L containers, capacity 0.33L per stroke. For 205L containers capacity 0.5L per stroke. Available in two sizes. With a 900mm flexible dispensing tube, except polypropylene 205L dispensing pump.
- Manufactured with nylon for maximum resistance to acids and solvents
- No metal part comes into contact with the liquid being pumped
- PTFE seals fitted
- Universal adapters fitted to provide secure seating in the necks of virtually all drums from 20L/25L to 205L capacities
- Piston action ensures that the liquid flow is totally under control at all times during the action For dispensing solvents and acids in controlled amounts.
Specifications
Specifications
| Quantity | 1 Each |
| Type | Lift Pump |
| For Use With (Application) | For dispensing solvents and acids in controlled amounts |
| Material | Nylon |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.