Description
- The lid has been designed allowing full use of whole width of a water bath with the lowest height of the gable being 45mm above the bath.
- This feature means test tube racks or flasks can be placed across the whole width of the bath without fouling the lid, maximising the bath working area.
- Designed with customer in mind ensuring where condensation accumulates in use there are no drip points which could spoil work in open flasks in the water bath.
- Water evaporation is reduced at elevated temperatures, maintaining water levels in the water bath.
- Where an accessory TC-1 Thermometer is used the temperature can be read from the thermometer with the lid in place.
- Lid fits into the tank ensuring all condensation runs back into the bath and not down the side onto your bench top.
- Reduces energy consumption and running costs for the water bath.
- Insulated handle.
Specifications
Specifications
| For Use With (Equipment) | 2 L and 4 L Unstirred Baths |
| Type | Lift Off Gable Lid |