Learn More
Lifitegrast, MedChemExpress
Product Code. 16497738
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16497738 10 mg 10mg
16433717 50 mg 50mg
16407606 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16497738 Supplier Medchem Express Supplier No. HY19344
 Restricted Item
Estimated Shipment: 06-10-2026
MedChemExpress Lifitegrast (SAR 1118) is a potent integrin antagonist. Lifitegrast blocks the binding of intercellular adhesion molecule 1 (ICAM-1) to lymphocyte function-associated antigen 1 (LFA-1), interrupting the T cell-mediated inflammatory cycle. Lifitegrast inhibits Jurkat T cell attachment to ICAM-1 with an IC50 of 2.98 nM. Lifitegrast can be used for researching dry eye disease.
Lifitegrast (SAR 1118) is a potent integrin antagonist. Lifitegrast blocks the binding of intercellular adhesion molecule 1 (ICAM-1) to lymphocyte function-associated antigen 1 (LFA-1), interrupting the T cell-mediated inflammatory cycle. Lifitegrast inhibits Jurkat T cell attachment to ICAM-1 with an IC50 of 2.98 nM. Lifitegrast can be used for researching dry eye disease.
Specifications
Chemical Name or Material Lifitegrast
CAS 1025967-78-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C29H24Cl2N2O7S
Quantity 10 mg
Synonym SAR 1118 SHP-606
Solubility Information DMSO : ≥ 29 mg/mL (47.12 mM)
SMILES O=C(O)[C@H](CC1=CC=CC(S(=O)(C)=O)=C1)NC(C2=C(Cl)C3=C(CN(C(C4=CC=C5C=COC5=C4)=O)CC3)C=C2Cl)=O
Molecular Weight (g/mol) 615.48
Formula Weight 615.48
Percent Purity 99.09%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation