Learn More
Applied Biosystems™ Region of Interest (ROI) and Background Plates, Fast 96-well
Product Code. 12323583 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 x 96-well Plate
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12323583 1 x 96-well Plate Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12323583 Supplier Applied Biosystems™ Supplier No. 4432426
Black Friday Starts Here
Use promo code 27984
View all promotions
 Restricted Item
Estimated Shipment: 08-01-2027
Add to basket
Edge
Add to basket
Used to maintain the ViiA™ 7, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6, QuantStudio™ 7, and QuantStudio™ 12K Flex real-time PCR systems with Fast 96-well (0.1mL) blocks.
Calibration and verification should be run at least every six months. Please see instrument maintenance guide for full instructions.
Order Info
Dry ice shipping; store plates at -15° to -25°C
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at -15°C to -25°C
For Use With (Equipment) ViiA™ 7 System
Product Type Background Plate
For Use With (Application) Real Time PCR (qPCR)
Shipping Condition Dry Ice
Quantity 1 x 96-well Plate
Calibration Type Spectral Calibration