Learn More
Invitrogen™ E-Gel™ Imager Universal Filter
Description
Includes
1 x E-Gel Imager Universal Filter
E-Gel Imager is a very powerful and flexible system that allows to document gel images using multiple nucleic acid dyes and even Qdot™ nanocrystals. Each E-Gel Imager system includes a universal filter along with the camera hood.
- Change filters easily without needing any tools
- Filter Trays Allow Easy Swapping with No Extra Tools
- Each E-Gel Imager Camera Hood comes with a pre-installed universal filter
- Alternative filters can be easily utilized by simply removing the universal filter tray and sliding in the filter tray of your choosing into the camera hood
- Right Filter for most applications
- Universal filter installed in the camera hood at the time of purchase will be acceptable
- Label or Dye: Ethidium bromide
Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting
Specifications
Specifications
| Content And Storage | 1 x E-Gel™ Imager Universal Filter. Store at room temperature. |
| For Use With (Equipment) | E-Gel Imager |
| Product Line | E-Gel |
| Type | Imager Filter |
| Filter Type | Emission Filters |
| Label or Dye | Ethidium bromide, SYBR™ Safe |
| Shipping Condition | Room Temperature |
For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.