Learn More
Invitrogen™ Coverglass Removal Tool
Product Code. 15499814
Change view
Click to view available options
Quantity:
1 each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15499814 1 each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15499814 Supplier Invitrogen™ Supplier No. C37010
In Stock
Minimize the risk of breakage of chambered coverglass during cell culture and fluorescence imaging applications. CultureWell&Trade; Coverglass Removal Tool enables the chambered upper structure to be easily separated from the coverglass without the need for excessive force.
The Coverglass Removal Tool is intended for use with a CultureWell™ Chambered Coverglass for Cell Culture (Mfc. Nos. C37000, C37005). The removable chambered coverglass is provided sterile and ready to use. It contains 16 wells that can each hold up to 250μL, allowing cells to be cultured in a number of different conditions on a single slide. When the cells are ready to be imaged, coverglass removal is made easy by the use of this simple tool.
TRUSTED_SUSTAINABILITY
Specifications
Type Coverglass Removal Tool
Product Line Culturewell
For Use With (Equipment) Microscope
Quantity 1 each