Learn More
Applied Biosystems™ Calibration Kit, Fast 96-well
Product Code. 15810403 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 x 96-well plate
Unit Size:
Each
Product Code. Quantity unitSize
15810403 1 x 96-well plate Each
1 options
Product Code. 15810403
Brand: Applied Biosystems™ 4432563
Includes
Region of interest (ROI) plate, Background plate, Dye calibration plates, and RNase P instrument verification plate
Used to maintain your real-time PCR instrument with Fast 96-well block.
The kit includes region of interest (ROI), background, and dye calibrations, as well as RNase P verification. Please see your instrument user guide for full instructions on use.
See instrument user guide for full instructions.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Region-of-Insert (ROI)
Calibrated Dye Proprietary Mix
For Use With (Equipment) ViiA™ 7 Dx System, QuantStudio™ Dx
Product Type Calibration Kit
For Use With (Application) Real Time PCR (qPCR)
Quantity 1 x 96-well plate
Calibration Type Spectral Calibration
For in vitro Diagnostic Use.