Learn More
BRAND™ Life Science Sealing Films for PCR and Storage
Shop All Brand Products
Click to view available options
Material:
Aluminum
Polyester
Length (Metric):
16.5 cm
17.2 cm
Unit Size:
Pack of 100
Description
- Due to its adhesive, film can be used down to -80°C
- DMSO-resistant and resistant to common solvents
- Can be removed without leaving any residue so that work can continue unhindered after transport or storage
Specifications
Specifications
| Color | Silver |
| Description | Sealing film, aluminum, peelable |
| Material | Aluminum |
| Sterility | Non-sterile |
| Autoclavable | Not Autoclavable |
| Length (Metric) | 16.5 cm |
| Width (Metric) | 9 cm |
| Pierceable | No |
| Temperature Range (Metric) | -80°C to +120°C |
| Fit for Purpose | PCR Ready |