Learn More
Karl Hecht Liebig cooler with cut, DURAN 25cm
Brand:  Karl Hecht 41982040
Liebig cooler with cut, DURAN 25cm
Product Code. 10013951
  • ONLINE EXCLUSIVE PRICE
    787.50 NOK
    584.55 NOK / Each
    Save 202.95 NOK
    25% Off
Only null left
Estimated Shipment: 27-02-2026