Learn More
Haldenwanger™ Lids for Porcelain Crucibles
Use these lids with porcelain crucibles. Haldenwanger™ Lids for Porcelain Crucibles are constructed from optimized laboratory porcelain.
87.55 NOK - 959.30 NOK
Product Code | Brand | Diameter (Metric) Outer | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outer | Price | Quantity & Availability | |||||
12376487
|
Haldenwanger™
79D/8 |
32mm |
757.90 NOK
Pack of 10 |
Estimated Shipment: 28-06-2024
Log in to see stock. |
|||||
12356507
|
Haldenwanger™
79D/7 |
35mm |
650.00 NOK
Pack of 10 |
Estimated Shipment: 29-04-2024
Log in to see stock. |
|||||
12306497
|
Haldenwanger™
79D/4 |
60mm |
577.70 NOK
Pack of 5 |
Estimated Shipment: 29-04-2024
Log in to see stock. |
|||||
12366507
|
Haldenwanger™
L 79D/5 |
52mm |
959.30 NOK
Pack of 10 |
Estimated Shipment: 10-06-2024
Log in to see stock. |
|||||
12867713
|
Haldenwanger™
79D/3 |
68mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12346507
|
Haldenwanger™
79D/9 |
28mm |
657.20 NOK
Pack of 10 |
Estimated Shipment: 29-04-2024
Log in to see stock. |
|||||
10353121
|
Haldenwanger™
L 79D/5 |
52mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Available in a variety of sizes
- For use with Haldenwanger porcelain crucibles
- Complies with DIN VDE 0335, type C 100
Specifications
Porcelain | |
Crucible Lid |