Learn More
Haldenwanger™ Lids for Porcelain Crucibles
Product Code. 12867713
Change view
Click to view available options
Diameter (Metric) Outer:
28mm
32mm
35mm
52mm
60mm
68mm
Unit Size:
Each
Pack of 10
Pack of 5
Product Code. Diameter (Metric) Outer unitSize
12867713 68mm Pack of 5
12346507 28mm Pack of 10
12376487 32mm Pack of 10
12356507 35mm Pack of 10
12306497 60mm Pack of 5
12366507 52mm Pack of 10
10353121 52mm Each
7 options
Product Code. 12867713
Brand: Haldenwanger™ 79D/3
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use these lids with porcelain crucibles. Haldenwanger™ Lids for Porcelain Crucibles are constructed from optimized laboratory porcelain.
  • Available in a variety of sizes
  • For use with Haldenwanger porcelain crucibles
  • Complies with DIN VDE 0335, type C 100
Specifications
Material Porcelain
Diameter (Metric) Outer 68mm
Product Type Crucible Lid