missing translation for 'onlineSavingsMsg'
Learn More

Lidocaine Hydrochloride, MP Biomedicals

Product Code. 11476321 Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 g
250 g
Unit Size:
10g
250g
This item is not returnable. View return policy

Product Code. 11476321

Brand: MP Biomedicals 0219391783

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 6108-05-0
Molecular Formula C14H25ClN2O2
Molecular Weight (g/mol) 288.82
MDL Number MFCD00150329
InChI Key YECIFGHRMFEPJK-UHFFFAOYSA-N
Synonym lidocaine hydrochloride monohydrate, 2-diethylamino-n-2,6-dimethylphenyl acetamide hydrochloride hydrate, dolicaine, linocaine hydrochloride, lidocaine hydrochloride hydrate, xylocaine hydrochloride monohydrate, lignocaine hydrochloride monohydrate, 2-diethylamino-n-2,6-dimethylphenyl acetamide hydrochloride monohydrate, zingo tm, lidocaine hcl hydrate
PubChem CID 16219577
ChEBI CHEBI:60791
SMILES O.[Cl-].CC[NH+](CC)CC(=O)NC1=C(C)C=CC=C1C

Specifications

Quantity 250 g
IUPAC Name {[(2,6-dimethylphenyl)carbamoyl]methyl}diethylazanium hydrate chloride
Formula Weight 270.8
Chemical Name or Material Lidocaine Hydrochloride
compliance-icons
Product Identifier
  • Lidocaine Hydrochloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.