Learn More
Buerkle™ Lid for Round Containers
Product Code. 17998441 Shop All Bürkle Products
Change view
Click to view available options
Diameter (English) Outer:
9.65 in.
18.5 in.
21.65 in.
27.17 in.
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (English) Outer Unit Size
17998441 9.65 in. Each
17860974 18.5 in. Each
17880974 21.65 in. Each
17806414 27.17 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 17998441 Supplier Buerkle™ Supplier No. 34900141
  • ONLINE EXCLUSIVE PRICE
    1315.00 NOK
    1191.37 NOK / Pack of 1
    Save 123.63 NOK
    9% Off
Estimated Shipment: 05-01-2027
Lid for Buerkle round shaped containers.
  • L-LDPE (linear LDPE), transparent
  • Rupture-proof
TRUSTED_SUSTAINABILITY
Specifications
Diameter (English) Outer 9.65 in.
Type Tank Lid