Learn More
Labconco™ Lid for FreeZone Freeze Dryer
Product Code. 17281134 Shop All Labconco Corporation Products
Change view
Click to view available options
For Use With (Equipment):
FreeZone Freeze Dryer
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
17281134 FreeZone Freeze Dryer Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17281134 Supplier Labconco™ Supplier No. 7532900
  • ONLINE EXCLUSIVE PRICE
    1535.00 NOK
    1469.10 NOK / Pack of 1
    Save 65.90 NOK
    4% Off
 Restricted Item
Estimated Shipment: 28-01-2027
Replacement lid for FreeZone Freeze Dryer
  • Replacement lid for FreeZone Freeze Dryer.
  • 3/4 in. lid, 14 in. diameter lid fits on drying chambers and FreeZone collector chambers.
Specifications
Type Lid
Capacity (Metric) Up to 18 L
Temperature Range (Metric) -105°C to +170°C
Material Acrylic
For Use With (Equipment) FreeZone Freeze Dryer
Height (English) 0.75 in.
Height (Metric) 19.05 mm
Length (English) 14 in.
Length (Metric) 355.6 mm