missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lgr6 Antibody (918726) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB84581JF525
This item is not returnable.
View return policy
Description
Lgr6 Monoclonal antibody specifically detects Lgr6 in Human samples. It is validated for Flow Cytometry,Blockade of Receptor-ligand Interaction,Immunocytochemistry,CyTOF
Specifications
| Lgr6 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| FLJ14471, gonadotropin receptor, GPCR, leucine-rich repeat containing G protein-coupled receptor 6, leucine-rich repeat-containing G protein-coupled receptor 6, leucine-rich repeat-containing G-protein coupled receptor 6, VTS20631 | |
| NS0 mouse myeloma cell line transfected with human Lgr6, Met1-Val967, Accession # Q9HBX8 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, Blockade of Receptor-ligand Interaction, Immunocytochemistry, CyTOF | |
| 918726 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| GPCR | |
| 59352 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction