Learn More
LGR4 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Product Code. 16889654 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16889654 150 μL 150µL
16899654 20 μL 20µL
2 options
Product Code. 16889654
Brand: Proteintech 201501AP150UL
Rabbit Polyclonal Antibody
The LGR4 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human LGR4. This antibody recognizes human, mouse, rat antigen. The LGR4 antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
Antigen LGR4
Applications Western Blot, Immunohistochemistry, ELISA
Classification Polyclonal
Concentration 227 μg/mL
Conjugate Unconjugated
Host Species Rabbit
Quantity 150 μL
Primary or Secondary Primary
Gene ID (Entrez) 55366
Target Species Human, Rat, Mouse
Isotype IgG