Learn More
LGMN Mouse anti-Human, Unconjugated, Clone: 1F2B12, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67017-1-IG-150UL
Additional Details : Weight : 0.00010kg
Description
The LGMN antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human LGMN. This antibody recognizes human antigen. The LGMN antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
LGMN | |
Monoclonal | |
960 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, Immunohistochemistry, ELISA | |
1F2B12 | |
Unconjugated | |
150 μL | |
5641 | |
IgG1 |