missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LGI2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18044-1-AP-150UL
This item is not returnable.
View return policy
Description
The leucine-rich, glioma inactivated gene 2 (LGI2) is a member of the LGI family in which LGI1 is the exemplar. The LGI family consists of four of highly related proteins containing leucine-rich repeats (LRRs) which are highly similar to other transmembrane signaling molecules and receptors. LGI1 has been identified as a candidate tumor suppressor gene for glioma and plays a role in autodomit lateral temporal epilepsy (ADTLE), an epileptic syndrome characterized by focal seizures with predomit auditory symptoms. Despite its high homology with LGI1 and similar pattern of expression, mutations in LGI2 have not been found to be associated with ADTLE.Specifications
| LGI2 | |
| Polyclonal | |
| Unconjugated | |
| LGI2 | |
| KIAA1916, LGI1 like protein 2, LGI2, LGIL2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 246316, 305417, 55203 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.31 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8K4Z0, Q8N0V4 | |
| LGI2 | |
| LGI2 Fusion Protein Ag12680 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction