Learn More
LGD 3303, TRC
Specifications
Specifications
| Chemical Name or Material | 9-chloro-2-ethyl-1-methyl-3-(2,2,2-trifluoroethyl)-6H-pyrrolo[3,2-f]quinolin-7-one |
| CAS | 917891-35-1 |
| Molecular Formula | C16 H14 Cl F3 N2 O |
| Quantity | 5 mg |
| Synonym | 9-Chloro-2-ethyl-3,6-dihydro-1-methyl-3-(2,2,2-trifluoroethyl)-7H-pyrrolo[3,2-f]quinolin-7-one, LGD 3303 |
| InChI Formula | InChI=1S/C16H14ClF3N2O/c1-3-11-8(2)14-12(22(11)7-16(18,19)20)5-4-10-15(14)9(17)6-13(23)21-10/h4-6H,3,7H2,1-2H3,(H,21,23) |
| SMILES | CCc1c(C)c2c3C(=CC(=O)Nc3ccc2n1CC(F)(F)F)Cl |
| IUPAC Name | 9-chloro-2-ethyl-1-methyl-3-(2,2,2-trifluoroethyl)-6H-pyrrolo[3,2-f]quinolin-7-one |
| Molecular Weight (g/mol) | 342.74 |
| Formula Weight | 342.0747 |