missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ LGALS1 (Human) Recombinant Protein
Human LGALS1 full-length ORF ( AAH01693, 1 a.a. - 135 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00003956-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MACGLVASNLNLKPGECLRVRGEVAPDAKSFVLNLGKDSNNLCLHFNPRFNAHGDANTIVCNSKDGGAWGTEQREAVFPFQPGSVAEVCITFDQANLTVKLPDGYEFKFPNRLNLEAINYMAADGDFKIKCVAFD
Specifications
AAH01693 | |
lectin, galactoside-binding, soluble, 1 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
LGALS1 | |
GST |
3956 | |
Wheat germ expression system | |
25 μg | |
DKFZp686E23103, GAL1, GBP | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |