Learn More
Thermo Scientific Chemicals Leuprolide
Product Code. 15464569
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
Product Code. Quantity unitSize
15464569 1 mg 1mg
15454569 5 mg 5mg
2 options
Product Code. 15464569
Brand: Thermo Scientific Chemicals J62147.M¬
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For cellular and molecular biology applications
Highly active luteinizing hormone releasing hormone (LHRH) agonist
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Purification Method HPLC
Common Name Leuprorelin
Format Powder
For Use With (Application) Highly active luteinizing hormone releasing hormone (LHRH) agonist
Molecular Weight (g/mol) 1209.41
Product Type Leuprolide
Quantity 1 mg
Regulatory Status RUO - research use only
Sequence [Des-Gly10, D-Leu6, Pro-NHEt9]LH-RH