Learn More
Thermo Scientific Chemicals Leuprolide
Description
Highly active luteinizing hormone releasing hormone (LHRH) agonist
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Purification Method | HPLC |
| Common Name | Leuprorelin |
| Format | Powder |
| For Use With (Application) | Highly active luteinizing hormone releasing hormone (LHRH) agonist |
| Molecular Weight (g/mol) | 1209.41 |
| Product Type | Leuprolide |
| Quantity | 5 mg |
| Regulatory Status | RUO - research use only |
| Sequence | [Des-Gly10, D-Leu6, Pro-NHEt9]LH-RH |