Learn More
Leupeptin hemisulfate, MedChemExpress
Product Code. p-200098838
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Product Code. 16439967
Brand: Medchem Express HY18234A5mg
MedChemExpress Leupeptin hemisulfate is a membrane-permeable thiol protease inhibitor that inhibits Cathepsin B, Cathepsin H and Cathepsin L, and also impairs amphisome-lysosome fusion. Leupeptin hemisulfate also exhibits anti-inflammatory effect.
Leupeptin hemisulfate is a membrane-permeable thiol protease inhibitor that inhibits Cathepsin B, Cathepsin H and Cathepsin L, and also impairs amphisome-lysosome fusion. Leupeptin hemisulfate also exhibits anti-inflammatory effect.
Specifications
Chemical Name or Material Leupeptin hemisulfate
Physical Form Powder
CAS 103476-89-7
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C20H38N6O4.1/2H2SO4
For Use With (Application) COVID-19-immunoregulation
Solubility Information H2O : 83.33 mg/mL (175.21 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C[C@@H](NC([C@@H](NC([C@@H](NC(C)=O)CC(C)C)=O)CC(C)C)=O)CCCNC(N)=N.[0.5H2SO4]
Molecular Weight (g/mol) 475.59
Formula Weight 475.59
Percent Purity 98.0%
Grade Research
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.