missing translation for 'onlineSavingsMsg'
Learn More

LETM1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16855474 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16855474 20 μL 20µL
16845474 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 16855474

Brand: Proteintech 160241AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This gene encodes a protein that is localized to the inner mitochondrial membrane. The protein functions to maintain the mitochondrial tubular shapes and is required for normal mitochondrial morphology and cellular viability. Mutations in this gene cause Wolf-Hirschhorn syndrome, a complex malformation syndrome caused by the deletion of parts of the distal short arm of chromosome 4. Related pseudogenes have been identified on chromosomes 8, 15 and 19.

Specifications

Antigen LETM1
Applications Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 0.35 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene LETM1
Gene Accession No. O95202, Q5XIN6, Q9Z2I0
Gene Alias LETM1
Gene Symbols Letm1
Host Species Rabbit
Immunogen LETM1 Fusion Protein Ag8861
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 305457, 3954, 56384
Target Species Rat, Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.