missing translation for 'onlineSavingsMsg'
Learn More

Letermovir, MedChemExpress

Product Code. p-200101126
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16422497

Brand: Medchem Express HY15233

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Letermovir (AIC246) is a potent inhibitor of CMV, which targets the viral terminase complex and remains active against virus resistant to DNA polymerase inhibitors.

Letermovir (AIC246) is a potent inhibitor of CMV, which targets the viral terminase complex and remains active against virus resistant to DNA polymerase inhibitors.

Specifications

Chemical Name or Material Letermovir
CAS 917389-32-3
Color Yellow
Purity Grade Notes Research
Molecular Formula C29H28F4N4O4
Quantity 50 mg
Synonym AIC246
Solubility Information DMSO : ≥ 100 mg/mL (174.66 mM)
SMILES FC1=C(N=C(N2CCN(C3=CC=CC(OC)=C3)CC2)N(C4=CC(C(F)(F)F)=CC=C4OC)[C@H]5CC(O)=O)C5=CC=C1
Molecular Weight (g/mol) 572.55
Formula Weight 572.55
Percent Purity 98.67%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.