Learn More
Letermovir, MedChemExpress
Product Code. 16422497
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16422497
Brand: Medchem Express HY15233
MedChemExpress Letermovir (AIC246) is a potent inhibitor of CMV, which targets the viral terminase complex and remains active against virus resistant to DNA polymerase inhibitors.
Letermovir (AIC246) is a potent inhibitor of CMV, which targets the viral terminase complex and remains active against virus resistant to DNA polymerase inhibitors.
Specifications
Chemical Name or Material Letermovir
CAS 917389-32-3
Color Yellow
Purity Grade Notes Research
Molecular Formula C29H28F4N4O4
Quantity 50 mg
Synonym AIC246
Solubility Information DMSO : ≥ 100 mg/mL (174.66 mM)
SMILES FC1=C(N=C(N2CCN(C3=CC=CC(OC)=C3)CC2)N(C4=CC(C(F)(F)F)=CC=C4OC)[C@H]5CC(O)=O)C5=CC=C1
Molecular Weight (g/mol) 572.55
Formula Weight 572.55
Percent Purity 98.67%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.