Learn More
Leteprinim Potassium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | Leteprinim Potassium Salt |
| CAS | 192564-13-9 |
| Molecular Formula | C15 H12 N5 O4 . K |
| Quantity | 10 mg |
| InChI Formula | InChI=1S/C15H13N5O4.K/c21-11(19-10-3-1-9(2-4-10)15(23)24)5-6-20-8-18-12-13(20)16-7-17-14(12)22;/h1-4,7-8H,5-6H2,(H,19,21)(H,23,24)(H,16,17,22);/q;+1/p-1 |
| SMILES | [K+].[O-]C(=O)c1ccc(NC(=O)CCn2cnc3C(=O)NC=Nc23)cc1 |
| IUPAC Name | potassium;4-[3-(6-oxo-1H-purin-9-yl)propanoylamino]benzoate |
| Molecular Weight (g/mol) | 365.39 |
| Formula Weight | 365.0526 |