Learn More
Leniolisib, MedChemExpress
Product Code. 16497406
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
Product Code. Quantity unitSize
16497406 100 mg 100mg
16457438 10 mg 10mg
16439917 5 mg 5mg
16431719 1 mg 1mg
16403467 50 mg 50mg
5 options
Product Code. 16497406
Brand: Medchem Express HY17635
MedChemExpress Leniolisib (CDZ173) is a potent and selective PI3Kδ inhibitor. Leniolisib has the potential for immunodeficiency disorders treatment.
Leniolisib (CDZ173) is a potent and selective PI3Kδ inhibitor. Leniolisib has the potential for immunodeficiency disorders treatment.
Specifications
Chemical Name or Material Leniolisib
CAS 1354690-24-6
Color White
Purity Grade Notes Research
Molecular Formula C21H25F3N6O2
Quantity 100 mg
Synonym CDZ173
Solubility Information DMSO : 100 mg/mL (222.00 mM; Need ultrasonic)
SMILES CCC(N1C[C@@H](NC2=C(CN(C3=CC(C(F)(F)F)=C(OC)N=C3)CC4)C4=NC=N2)CC1)=O
Molecular Weight (g/mol) 450.46
Formula Weight 450.46
Percent Purity 98.29%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation