Learn More
Lemildipine, MedChemExpress
Product Code. 16450308
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Product Code. 16450308
Brand: Medchem Express HY19663
MedChemExpress Lemildipine is a new dihydropyridine calcium entry blocker.
Lemildipine is a new dihydropyridine calcium entry blocker.
Specifications
Chemical Name or Material Lemildipine
CAS 94739-29-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C20H22Cl2N2O6
Quantity 5 mg
Synonym NB-818 NPK-1886
Solubility Information DMSO : 100 mg/mL (218.67 mM; Need ultrasonic)
SMILES O=C(C1=C(COC(N)=O)NC(C)=C(C(OC)=O)C1C2=CC=CC(Cl)=C2Cl)OC(C)C
Molecular Weight (g/mol) 457.3
Formula Weight 457.3
Percent Purity 98.06%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.