missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Leishmania LPG Monoclonal Antibody (CA7AE)

Description
Reconstitute with 0.5 mL of distilled water. Prior to reconstitution, store at 4ºC. After reconstitution, store undiluted at -20ºC, avoiding freeze/thaw cycles. Mouse anti Leishmania lipophosphoglycan antibody, clone CA7AE recognizes lipophosphoglycan (LPG) the major cell surface glycoconjugate of Leishmania parasites.
Lipophosphoglycan (LPG) is the major cell surface glycoconjugate of Leishmania parasites. LPG antibodies recognize the repeat carbohydrate epitope of most species of Leishmania. The epitope is also found on the excreted acid phosphatase of Leishmania and is expressed on the surface of Leishmania infected macrophages (Tolson et al. 1990).
Specifications
Specifications
| Antigen | Leishmania LPG |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | CA7AE |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | ascites with no preservative |
| Gene Alias | L. pneumophila; Legionellosis |
| Host Species | Mouse |
| Immunogen | Heat killed L. donovani promastigotes. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?