Learn More
Leishman'S Stain, Pure, Fisher Chemical
Product Code. 10122150 Shop All Fisher Chemical Products
Click to view available options
Quantity:
10 g
Unit Size:
10g
Product Code. 10122150
Brand: Fisher Chemical L/1815/45
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
CAS 12627-53-1
Color Dark green
Format Powder Solid
Formula Weight 171.0387g/mol
Melting Point 79°C/174.2°F
Packaging Glass Jar
Quantity 10 g
compliance-icons
Product Identifier
  • Leishman stain
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P311-Call a POISON CENTER/doctor/ .
  • P362-Take off contaminated clothing.