Learn More
Thermo Scientific™ Legend XFR TX-1000 Cell Culture Centrifuge Package
Product Code. 15374406
Change view
Click to view available options
:
Legend XFR TX-1000 Cell Culture Centrifuge Package
Product Code.
15374406 Legend XFR TX-1000 Cell Culture Centrifuge Package
1 options
Product Code. 15374406
Brand: Thermo Scientific™ 50146602
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Includes
Sorvall Legend XFR, refrigerated, 230V, 50/60Hz (Mfr PN: 75004538); TX-1000 rotor, Slide coat (Mfr PN: 75003017); TX-1000 buckets (set of 4) (Mfr PN: 75003001); TX-1000 ClickSeal biocontainment lids (set of 4) (Mfr PN: 75007309); Adapter, 50mL Conical (Mfr PN: 75003674); Adapter, 24 x 15mL, BIOFlex HC (Mfr PN: 75007306)
Process up to 40% more conical tubes in swing-out rotor and 100% more in fixed angle configuration.
Delivers high-performance processing with unparalleled processing performance, maximum versatility, and exceptional energy efficiency.
TRUSTED_SUSTAINABILITY
Specifications
Description Legend XFR TX-1000 Cell Culture Centrifuge Package
Includes Sorvall Legend XFR, refrigerated, 230V, 50/60Hz (Mfr PN: 75004538); TX-1000 rotor, Slide coat (Mfr PN: 75003017); TX-1000 buckets (set of 4) (Mfr PN: 75003001); TX-1000 ClickSeal biocontainment lids (set of 4) (Mfr PN: 75007309); Adapter, 50mL Conical (Mfr PN: 75003674); Adapter, 24 x 15mL, BIOFlex HC (Mfr PN: 75007306)