Learn More
Motic™ LED Module, AE2000/AE31
Product Code. 11829113
Change view
Click to view available options
For Use With (Equipment):
AE2000/AE31 Microscopes
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
11829113 AE2000/AE31 Microscopes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11829113 Supplier Motic™ Supplier No. 1101000201561
Only null left
Add to basket
Add to basket
Long-lasting transmitted light source for microscope illumination. Motic™ LED Module, AE2000/AE31 has a 6000°K color temperature.
  • AE2000/AE31 Microscope lighting module.
  • Long-lasting LED.
  • 6000°K color temperature
Specifications
Type LED Transmitted Light Module
Description 6000°K ±300°K
For Use With (Equipment) AE2000/AE31 Microscopes