missing translation for 'onlineSavingsMsg'
Learn More

Lectin, Wheat Germ Agglutinin, MP Biomedicals™

Product Code. 11418511 Shop All MP Biomedicals Products
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
This item is not returnable. View return policy

Product Code. 11418511

Brand: MP Biomedicals 0215226605

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Valuable in a variety of applications, including mitogenic stimulation of lymphocytes. Lectin, Wheat Germ Agglutinin, MP Biomedicals is not blood group specific, but has an affinity for N-acetyl-β-D-glucosamine residues and oligomers. WGA contains no

Lectins are proteins or glycoproteins of non-immune origin that agglutinate cells and/or precipitate complex carbohydrates. Lectins are capable of binding glycoproteins even in presence of various detergents. The agglutination activity of these highly specific carbohydrate-binding molecules is usually inhibited by a simple monosaccharide, but for some lectins, di, tri, and even polysaccharides are required.

The lecithin in this product is purified Triticum vulgrislectin from Wheat Germ Agglutinin (WGA). It is very valuable in a wide variety of applications in vitro, including:

  • Blood grouping and erythrocyte polyagglutination studies
  • Mitogenic stimulation of lymphocytes
  • Lymphocyte subpopulation studies
  • Fractionation of cells and other particles
  • Histochemical studies of normal and pathological conditions
TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 mg
Synonym WGA, Wheat germ agglutinin, Lectin from Triticum vulgaris (wheat)
Solubility Information Dissolvable in distilled water and 0.9% NaCl within 5 minutes.
Concentration or Composition (by Analyte or Components) Working Concentration: Reconstitute with Buffer to a concentration of 1mg/mL.
Physical Form Lyophilized Powder
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.