Learn More
Abeba™ Leather Working Shoes
Product Code. 15326656
Change view
Click to view available options
Size:
35
37
38
40
41
45
46
47
Unit Size:
1 pair
Product Code. Size unitSize
15326656 40 1 pair
15376646 35 1 pair
15396646 37 1 pair
15306656 38 1 pair
15336656 41 1 pair
15376656 45 1 pair
15386656 46 1 pair
15396656 47 1 pair
8 options
Product Code. 15326656
Brand: Abeba™ 7031/40
Only null left
Add to basket
Add to basket
Animal fats, oil and fuel resistant. Sanitized™ treated.
  • CE EN ISO 20347:2012 OB FO SRC
  • Smooth leather
  • Black
  • Slip resistance outsole sole
  • Sanitized treated
  • Heel strap foldable, adjustable
  • Leather insole
Specifications
Certifications/Compliance CE EN ISO 20347:2012 OB FO SRC
Color Black
Material Leather
Size 40
Sole Features Slip-resistant
Style 7031
Resistant To Animal fats, oil and fuel