missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead, Technical, SLR, Metal Powder, Below 200 Mesh, Fisher Chemical
Shop All Fisher Chemical Products
Click to view available options
Quantity:
500 g
Packaging:
Plastic powder jar
Unit Size:
500g
Specifications
Specifications
| Color | Gray |
| CAS | 7439-92-1 |
| Melting Point | 327.4°C |
| Boiling Point | 1740°C |
| Quantity | 500 g |
| Packaging | Plastic powder jar |
| Form | Solid |
| Molecular Formula | Pb |
| MDL Number | MFCD00134050 |
| Synonym | plumbum, metal, blei, lead, elemental, rough bullion, plomo, glover, element, flake, omaha & grant |
| Show More |
Safety and Handling
Product Identifier
- Lead
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Reproductive toxicity Category 1A
- Specific target organ toxicity after repeated exposure Category 2
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H360Df-May damage the unborn child. Suspected of damaging fertility.
- H373-May cause damage to organs.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction