Learn More
Lead solution for ICP/MS 10mg/L HNO3 matrix, Fisher chemical™
Product Code. 12967393 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
50 mL
100 mL
Packaging:
Bottle
Unit Size:
100mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
12967393 50 mL Bottle 50mL
12977393 100 mL Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
You may only order up to the available stock. For more details call customer services on +47 22 95 59 59.
Product Code. 12967393 Supplier Fisher Chemical Supplier No. S 641 050
  • ONLINE EXCLUSIVE PRICE
    1105.00 NOK
    796.92 NOK / 50mL
    Save 308.08 NOK
    27% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
You may only order up to the available stock. For more details call customer services on +47 22 95 59 59.
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Chemical Identifiers
CAS 7697-37-2
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O
Specifications
Packaging Bottle
Concentration 10 mg/L
Solvent or Matrix Nitric Acid
Quantity 50 mL
Chemical Name or Material Lead