Learn More
Lead(II) Iodide, Pure, SLR, Fisher Chemical
Product Code. 10624922 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glass Jar
Unit Size:
100g
Product Code. Quantity Packaging unitSize
10624922 100 g Glass Jar 100g
1 options
Product Code. 10624922
Brand: Fisher Chemical L/1400/48
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Melting Point 402°C
CAS 10101-63-0
Color Orange to Yellow
Physical Form Powder/Solid
Quantity 100 g
Molecular Formula I2Pb
MDL Number MFCD00011163
Synonym lead ii iodide, lead diiodide, plumbous iodide, lead iodide pbi2, pbi2, plumbum jodatum, blei ii-iodid, hsdb 636, diiodo-, e2-plumbane
InChI Key RQQRAHKHDFPBMC-UHFFFAOYSA-L
SMILES [I-].[I-].[Pb++]
IUPAC Name λ²-lead(2+) diiodide
Molecular Weight (g/mol) 461.00
PubChem CID 24931
Formula Weight 461g/mol
Packaging Glass Jar
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lead(II) iodide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.