Learn More
Kautex™ LDPE Narrow Neck Wash Bottle Closures
Product Code. 15961431
Change view
Click to view available options
For Use With (Equipment):
100 mL Bottle
1000 mL Bottle
200 mL Bottle
250 mL Bottle
500 mL Bottle
Diameter (Metric):
18 mm
25 mm
28 mm
Unit Size:
Each
Pack of 50
Product Code. For Use With (Equipment) Diameter (Metric) unitSize
15961431 500 mL Bottle 25 mm Pack of 50
15951431 200 mL Bottle 18 mm Pack of 50
15941431 100 mL Bottle 18 mm Pack of 50
10491151 100 mL Bottle 18 mm Each
10018330 250 mL Bottle 25 mm Each
10471151 500 mL Bottle 25 mm Each
10612421 1000 mL Bottle 28 mm Each
7 options
Product Code. 15961431
Brand: Kautex™ 2000075993
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Top off Series 301 and Series 312 round, narrow neck bottles with Kautex™ LPDE Narrow Neck Wash Bottle Closures. Their color is natural, which matches the bottle color.
  • LDPE
  • Natural color
  • Round
  • Wash bottle style
TRUSTED_SUSTAINABILITY
Specifications
Color Natural
Material LDPE
Product Type Wash Bottle Closure
For Use With (Equipment) 500 mL Bottle
Diameter (Metric) 25 mm