Learn More
Kautex™ LDPE Screw Closure
Product Code. 15991401
Change view
Click to view available options
Diameter (Metric):
25 mm
28 mm
40 mm
50 mm
65 mm
Unit Size:
Each
Pack of 100
Product Code. Diameter (Metric) unitSize
15991401 40 mm Pack of 100
15971401 25 mm Pack of 100
10674941 25 mm Each
10507721 28 mm Each
10527911 40 mm Pack of 100
15901411 50 mm Pack of 100
10353591 50 mm Each
10746791 65 mm Each
8 options
Product Code. 15991401
Brand: Kautex™ 2000070608
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Feature LDPE light weight, tough construction. Kautex™ LDPE Screw Closures are available in a wide variety of sizes for Kautex™ narrow and wide neck bottles.
  • Made from low density polyethylene
  • Translucent
  • Available for a wide variety of sizes
TRUSTED_SUSTAINABILITY
Specifications
Color Natural
Material LDPE
Thread Size (Metric) 00 mm
Neck Finish 00 mm
For Use With (Equipment) 200/250/300 mL Wide Mouth Bottle
Diameter (Metric) 40 mm