Description
- LDPE screw cap for narrow mouth bottles
- Fits GL 25 thread narrow neck laboratory bottles
- For storage of substances
- Includes 50 pack of screw caps
Specifications
Specifications
| Material | LDPE |
| Quantity | 50 Pk. |
| Thread Size (Metric) | GL 25 |
| Certified | Yes |
| Product Type | Screw Cap |
| For Use With (Equipment) | Fits GL 25 Thread narrow neck laboratory bottles |