missing translation for 'onlineSavingsMsg'
Learn More

Hirschmann™ LDPE Screw Cap, 10-pk

Click to view available options
Closure Size:
GL14
GL18
GL25
GL28
This item is not returnable. View return policy
This item is not returnable. View return policy

High quality screw cap for use with a variety of threaded bottles.

  • Different opening sizes can be achieved by cutting at any height
  • Natural/red color
  • With dropper nozzle and spout cape
Bottle Surface Transparent
Cleaning Procedure Gas/Chemical/Radiation
Color Natural, Red
Material LDPE
No. per Pack 10
Sterility Non-sterile
Thread Size (Metric) GL 28
Closure Color Natural, Red
Content And Storage Store at 5 to 40°C
Autoclavable Not Autoclavable
Closure Size GL28
Closure Material LDPE
Closure Type Screw Cap with Dropper Nozzle and Spout Cape
Shape Pointed
Show More Show Less