Learn More
Gilac™ Round Bucket with Stainless Steel Handle
Product Code. 11738511
Change view
Click to view available options
Color:
Blue
Red
White
Unit Size:
Each
Capacity (Metric):
13 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize Capacity (Metric)
11738511 White Each -
11722525 Red Each -
11742525 Blue Each 13 L
11752525 Red Each -
11748511 White Each -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11738511 Supplier Gilac™ Supplier No. G615021
  • ONLINE EXCLUSIVE PRICE
    274.05 NOK
    206.13 NOK / Pack of 1
    Save 67.92 NOK
    24% Off
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
Round Bucket with Stainless Steel Handle Food bucket and anti-wear skirt for increased product durability. Highly durable.
  • Food bucket
  • Highly durable
  • Anti-wear skirt for increased product durability
  • Made from low density polyethylene making it highly resistant
  • Stainless steel handle
  • External dimensions include handles
Specifications
Product Type Industrial Bucket
Graduations Yes
Material LDPE
Capacity (English) 2.6 gal.
Color White
Dimensions 310 x 280 x 280 mm
Handle Material Steel
Height (English) 11 in.
Height (Metric) 280 mm
Length (English) 12.2 in.
Length (Metric) 310 mm
Quantity 1 Ea.
Width (English) 11 in.