Learn More
Gilac™ Round Bucket with Stainless Steel Handle
Click to view available options
Color:
Blue
Red
White
Unit Size:
Each
Capacity (Metric):
13 L
Description
- Food bucket
- Highly durable
- Anti-wear skirt for increased product durability
- Made from low density polyethylene making it highly resistant
- Stainless steel handle
- External dimensions include handles
Specifications
Specifications
| Product Type | Industrial Bucket |
| Graduations | Yes |
| Material | LDPE |
| Capacity (English) | 2.6 gal. |
| Color | Red |
| Dimensions | 310 x 280 x 280 mm |
| Handle Material | Steel |
| Height (English) | 11 in. |
| Height (Metric) | 280 mm |
| Length (English) | 12.2 in. |
| Show More |